Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGEF40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278529.1 | NP_001265458.1 | 3 | 557 | ||
NM_001278530.1 | NP_001265459.1 | 3 | 557 | |||
NM_018071.4 | NP_060541.3 | 3 | 557 | |||
XM_005267844.2 | XP_005267901.1 | 3 | 593 | |||
XM_011536937.2 | XP_011535239.1 | 3 | 593 | |||
XM_011536938.2 | XP_011535240.1 | 3 | 595 | |||
XM_017021434.1 | XP_016876923.1 | 4 | 850 | |||
XM_017021435.1 | XP_016876924.1 | 3 | 594 | |||
GenBank mRNA | AK074057.1 | - | 3 | 502 | ||
BC142692.1 | - | 3 | 500 | 40148708 | ||
BC151212.1 | - | 3 | 500 | |||
BX248768.1 | - | 3 | 544 |