Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMBR1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300750.1 | NP_001287679.1 | 7 | 1071 | ||
NM_001300751.1 | NP_001287680.1 | 6 | 869 | |||
NM_018113.3 | NP_060583.2 | 6 | 869 | |||
XM_011538563.2 | XP_011536865.1 | 6 | 1023 | |||
XM_011538564.2 | XP_011536866.1 | 6 | 1396 | |||
XM_011538565.2 | XP_011536867.1 | 5 | 1645 | |||
XM_011538566.2 | XP_011536868.1 | 6 | 684 | |||
XM_017019626.1 | XP_016875115.1 | 6 | 1271 | |||
XM_017019627.1 | XP_016875116.1 | 7 | 1060 | |||
XM_017019628.1 | XP_016875117.1 | 6 | 858 | |||
XM_017019629.1 | XP_016875118.1 | 7 | 1915 | |||
XM_017019630.1 | XP_016875119.1 | 4 | 2896 | |||
XM_017019631.1 | XP_016875120.1 | 6 | 684 | |||
GenBank mRNA | AB033000.1 | - | 6 | 636 | ||
AF260728.1 | - | 6 | 828 | |||
AK001356.1 | - | 7 | 852 | |||
AK001651.1 | - | 6 | 819 | |||
AK093570.1 | - | 1 | 655 | |||
AK298026.1 | - | 5 | 812 | |||
AK310064.1 | - | 7 | 1005 | |||
AL137599.1 | - | 3 | 262 | |||
AL556633.3 | - | 6 | 833 | |||
AY358572.1 | - | 6 | 833 | |||
BC008337.1 | - | 6 | 799 | 3506347 | ||
BC015015.1 | - | 6 | 598 | 3910520 | ||
BC031550.1 | - | 6 | 630 | 5194023 | ||
KU178711.1 | - | 6 | 524 | |||
KU178712.1 | - | 5 | 507 | |||
KU178713.1 | - | 5 | 490 | |||
KU178714.1 | - | 6 | 524 | |||
KU178715.1 | - | 6 | 524 |