Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASUN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018164.2 | NP_060634.2 | 11 | 1636 | ||
XM_005253422.3 | XP_005253479.1 | 11 | 1595 | |||
XM_011520750.2 | XP_011519052.1 | 11 | 1187 | |||
XM_011520751.2 | XP_011519053.1 | 11 | 1613 | |||
XM_011520753.2 | XP_011519055.1 | 5 | 359 | |||
XM_017019632.1 | XP_016875121.1 | 11 | 1169 | |||
XM_017019633.1 | XP_016875122.1 | 10 | 1579 | |||
XM_017019634.1 | XP_016875123.1 | 10 | 1561 | |||
XM_017019635.1 | XP_016875124.1 | 11 | 1169 | |||
XM_017019637.1 | XP_016875126.1 | 9 | 799 | |||
GenBank mRNA | AF274950.1 | - | 3 | 218 | ||
AK001222.1 | - | 2 | 121 | |||
AK001492.1 | - | 11 | 1265 | |||
AK001499.1 | - | 11 | 1301 | |||
AK225336.1 | - | 11 | 1287 | |||
AK297948.1 | - | 9 | 976 | |||
AY211927.1 | - | 4 | 296 | |||
BC003081.2 | - | 11 | 1325 | 2989678 | ||
BC008368.1 | - | 11 | 1283 | 3355282 |