Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008744.1 | NP_001008744.1 | 15 | 1789 | ||
NM_018319.3 | NP_060789.2 | 16 | 2012 | |||
XM_005267847.2 | XP_005267904.1 | 16 | 2686 | |||
XM_005267848.2 | XP_005267905.1 | 15 | 2030 | |||
XM_006720197.3 | XP_006720260.1 | 16 | 2253 | |||
XM_006720198.3 | XP_006720261.1 | 15 | 1908 | |||
XM_006720199.1 | XP_006720262.1 | 15 | 1394 | |||
XM_011536942.2 | XP_011535244.1 | 15 | 2429 | |||
XM_011536943.2 | XP_011535245.1 | 15 | 1822 | |||
XM_017021439.1 | XP_016876928.1 | 16 | 1920 | |||
XM_017021442.1 | XP_016876931.1 | 13 | 1638 | |||
XM_017021443.1 | XP_016876932.1 | 13 | 1137 | |||
XM_017021444.1 | XP_016876933.1 | 13 | 1138 | |||
XM_017021446.1 | XP_016876935.1 | 14 | 1444 | |||
GenBank mRNA | AF182003.1 | - | 6 | 458 | ||
AK001952.1 | - | 15 | 1753 | |||
AK023514.1 | - | 5 | 375 | |||
AK093235.1 | - | 17 | 2820 | |||
AK315549.1 | - | 15 | 1737 | |||
AL832288.1 | - | 7 | 731 | |||
BC006083.1 | - | 18 | 2176 | |||
BC015474.1 | - | 16 | 1963 | 3900062 | ||
BC107877.1 | - | 17 | 2062 | 6597752 | ||
BX161451.1 | - | 15 | 1924 |