Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF415 SIRNA Assays ›
Gene Symbol:
ZNF415
Gene Name:
zinc finger protein 415
Gene Aliases:
ZfLp
Chromosome Location:
Chr.19: 53107879 - 53133077 on Build GRCh38
UniGene ID:
Hs.147765
Species:
Human
siRNA ID
HSS125036
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF415
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001136038.2
NP_001129510.2
4
1059
NM_001164309.1
NP_001157781.1
4
932
NM_018355.3
NP_060825.2
4
827
NR_028343.1
7
1188
XM_006723267.1
XP_006723330.1
6
1145
XM_006723268.1
XP_006723331.1
4
893
XM_006723272.2
XP_006723335.1
5
925
XM_006723273.2
XP_006723336.1
5
1009
XM_006723274.2
XP_006723337.1
5
1086
XM_006723277.2
XP_006723340.1
5
1020
XM_006723279.1
XP_006723342.1
5
1179
XM_006723280.2
XP_006723343.1
2
635
XM_006723281.2
XP_006723344.1
3
661
XM_006723282.1
XP_006723345.1
3
970
XM_011527100.1
XP_011525402.1
5
1169
XM_011527101.2
XP_011525403.1
5
1144
XM_011527103.1
XP_011525405.1
5
964
XM_011527104.1
XP_011525406.1
5
1258
XM_017026952.1
XP_016882441.1
5
972
XM_017026953.1
XP_016882442.1
6
1292
XM_017026954.1
XP_016882443.1
6
1169
XM_017026955.1
XP_016882444.1
6
1120
XM_017026956.1
XP_016882445.1
7
1270
XM_017026957.1
XP_016882446.1
6
1008
XM_017026958.1
XP_016882447.1
7
1324
XM_017026959.1
XP_016882448.1
6
1195
XM_017026960.1
XP_016882449.1
5
910
XM_017026961.1
XP_016882450.1
6
1356
XM_017026962.1
XP_016882451.1
6
1201
XM_017026963.1
XP_016882452.1
5
1097
XM_017026964.1
XP_016882453.1
5
1047
XM_017026965.1
XP_016882454.1
4
1061
XM_017026966.1
XP_016882455.1
5
1189
XM_017026967.1
XP_016882456.1
4
987
XM_017026968.1
XP_016882457.1
6
1285
XM_017026969.1
XP_016882458.1
2
706
XM_017026970.1
XP_016882459.1
6
1162
XM_017026971.1
XP_016882460.1
6
1381
XM_017026972.1
XP_016882461.1
4
963
XM_017026973.1
XP_016882462.1
5
1155
XM_017026974.1
XP_016882463.1
6
1234
GenBank mRNA
AK002053.1
-
4
799
AK095504.1
-
7
1160
AY283600.1
-
4
799
BC063880.1
-
5
1010
30388597
BC069668.1
-
5
1009
7262632
DQ925695.1
-
3
661
DQ925696.1
-
5
998
DQ925697.1
-
6
1008
DQ925698.1
-
5
1110
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0