Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TXLNG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168683.1 | NP_001162154.1 | 3, 4 | 520 | ||
NM_018360.2 | NP_060830.2 | 5, 6 | 916 | |||
XM_017029631.1 | XP_016885120.1 | 2, 3 | 890 | |||
GenBank mRNA | AK002071.1 | - | 5, 6 | 911 | ||
AY739713.1 | - | 3, 4 | 470 | |||
BC101572.1 | - | 5, 6 | 883 | 8069078 | ||
BC101576.1 | - | 5, 6 | 883 | 8069082 |