Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
CDMO and Clinical Trial Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Instrument Management
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FAM63A SIRNA Assays ›
Gene Symbol:
FAM63A
Gene Name:
family with sequence similarity 63 member A
Gene Aliases:
-
Chromosome Location:
Chr.1: 150996737 - 151008378 on Build GRCh38
UniGene ID:
Hs.743952
Species:
Human
siRNA ID
HSS125050
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FAM63A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001040217.2
NP_001035307.1
10
1699
NM_001163258.1
NP_001156730.1
11
1762
NM_001163259.1
NP_001156731.1
10
1664
NM_001163260.1
NP_001156732.1
10
1575
NM_001319998.1
NP_001306927.1
11
3079
NM_018379.4
NP_060849.2
11
2363
XM_005245321.3
XP_005245378.1
10
2340
XM_005245325.3
XP_005245382.1
10
1695
XM_005245327.3
XP_005245384.1
9
1536
XM_005245328.3
XP_005245385.1
9
1533
XM_011509757.2
XP_011508059.1
10
2313
XM_011509759.2
XP_011508061.1
9
1916
XM_017001770.1
XP_016857259.1
9
2593
XM_017001771.1
XP_016857260.1
9
2331
XM_017001772.1
XP_016857261.1
10
2075
XM_017001773.1
XP_016857262.1
10
2310
XM_017001774.1
XP_016857263.1
10
2048
XM_017001775.1
XP_016857264.1
9
2178
XM_017001776.1
XP_016857265.1
9
2151
XM_017001777.1
XP_016857266.1
10
1689
GenBank mRNA
AB037811.1
-
9
1548
AK002142.1
-
10
1425
AK125493.1
-
10
1399
AK125959.1
-
11
3079
AK301946.1
-
11
1762
AK303962.1
-
10
1352
BC032321.1
-
10
1839
5240620
BC075812.1
-
11
2350
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-567f868b4f-2g8tf:80/10.239.165.190:80.
git-commit: 96b0dd45ad55c22ec1de1c352dff25df4a817ab2
git-url: git@github.com:thermofisher/magellan-core.git
git-branch: origin/release/2.8.0-2024.05.19-1.0