Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040428.3 | NP_001035518.1 | 11 | 1820 | ||
NM_018418.4 | NP_060888.2 | 12 | 1916 | |||
XM_005267851.1 | XP_005267908.1 | 12 | 1890 | |||
XM_005267852.1 | XP_005267909.1 | 11 | 1819 | |||
XM_005267854.1 | XP_005267911.1 | 13 | 1952 | |||
XM_005267855.1 | XP_005267912.1 | 12 | 1885 | |||
XM_011536951.1 | XP_011535253.1 | 12 | 2084 | |||
XM_011536952.1 | XP_011535254.1 | 10 | 1702 | |||
XM_011536953.1 | XP_011535255.1 | 11 | 2023 | |||
XM_017021452.1 | XP_016876941.1 | 12 | 2081 | |||
XM_017021453.1 | XP_016876942.1 | 12 | 2050 | |||
XM_017021454.1 | XP_016876943.1 | 13 | 1949 | |||
XM_017021455.1 | XP_016876944.1 | 12 | 1882 | |||
XM_017021456.1 | XP_016876945.1 | 13 | 1895 | |||
XM_017021457.1 | XP_016876946.1 | 11 | 2020 | |||
GenBank mRNA | AF144487.3 | - | 12 | 1896 | ||
AF144488.2 | - | 11 | 1201 | |||
AK055864.1 | - | 11 | 3488 | |||
AK290631.1 | - | 11 | 1791 | |||
AL136604.1 | - | 11 | 1800 | |||
BC008656.1 | - | 7 | 1348 | |||
BC090875.1 | - | 12 | 2203 | 30383306 | ||
CR533563.1 | - | 11 | 1645 | |||
CR936777.1 | - | 12 | 1862 | |||
EU668353.1 | - | 12 | 1892 |