Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018460.3 | NP_060930.3 | 14 | 1510 | ||
XM_011511479.2 | XP_011509781.1 | 15 | 1542 | |||
XM_011511483.1 | XP_011509785.1 | 10 | 1140 | |||
XM_017004499.1 | XP_016859988.1 | 14 | 1471 | |||
XM_017004500.1 | XP_016859989.1 | 13 | 1337 | |||
GenBank mRNA | AF212222.1 | - | 14 | 1462 | ||
AF217507.1 | - | 14 | 1459 | |||
AF217522.1 | - | 14 | 1463 | |||
AK226014.1 | - | 14 | 1445 | |||
AY219338.1 | - | 14 | 1448 | |||
BC016701.2 | - | 14 | 1416 | 4108644 | ||
BC038976.1 | - | 14 | 1415 | 5553184 |