Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LANCL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018697.3 | NP_061167.1 | 4 | 1173 | ||
GenBank mRNA | AB035966.1 | - | 4 | 650 | ||
AF353942.1 | - | 4 | 650 | |||
AJ278245.1 | - | 4 | 781 | |||
AK095857.1 | - | 4 | 1173 | |||
AK225958.1 | - | 4 | 626 | |||
AK301747.1 | - | 4 | 579 | |||
AK313326.1 | - | 4 | 918 | |||
AL713665.2 | - | 4 | 565 | |||
BC070049.1 | - | 4 | 869 | 30343188 |