Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034023.1 | NP_001029195.1 | 3 | 291 | ||
NM_001034024.1 | NP_001029196.1 | 3 | 251 | |||
NM_019100.4 | NP_061973.1 | 2 | 472 | |||
XM_005271037.4 | XP_005271094.1 | 2 | 462 | |||
XM_005271039.1 | XP_005271096.1 | 3 | 362 | |||
XM_005271040.1 | XP_005271097.1 | 3 | 239 | |||
XM_011541784.2 | XP_011540086.1 | 3 | 296 | |||
XM_011541785.2 | XP_011540087.1 | 3 | 316 | |||
XM_017001807.1 | XP_016857296.1 | 3 | 347 | |||
XM_017001808.1 | XP_016857297.1 | 3 | 350 | |||
GenBank mRNA | AB037846.1 | - | 3 | 256 | ||
AF265228.1 | - | 2 | 171 | |||
AK021605.1 | - | 3 | 235 | |||
AK289366.1 | - | 3 | 274 | |||
AK293598.1 | - | 3 | 282 | |||
AK298793.1 | - | 3 | 264 | |||
AK300217.1 | - | 3 | 249 | |||
AK300372.1 | - | 3 | 241 | |||
AK300442.1 | - | 3 | 279 | |||
AL136657.1 | - | 3 | 253 | |||
AL137200.1 | - | 3 | 255 | |||
BC002855.2 | - | 3 | 240 | 3635724 | ||
BC008053.2 | - | 2 | 434 | 3834056 | ||
CA842195.1 | - | 3 | 283 | |||
CB123593.1 | - | 2 | 461 |