Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BAIAP2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018842.4 | NP_061330.2 | 12 | 1582 | ||
GenBank mRNA | AK027142.1 | - | 3 | 372 | ||
AK074419.1 | - | 3 | 176 | |||
BC013888.2 | - | 12 | 1535 | 3842949 |