Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021156.3 | NP_066979.2 | 8 | 916 | ||
GenBank mRNA | AK075404.1 | - | 8 | 895 | ||
AK226005.1 | - | 7 | 450 | |||
AL118494.1 | - | 7 | 441 | |||
AY359004.1 | - | 8 | 833 | |||
BC033787.1 | - | 8 | 727 | 5165952 | ||
BC044777.1 | - | 8 | 916 |