Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAR1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142648.1 | NP_001136120.1 | 4 | 363 | ||
NM_020150.4 | NP_064535.1 | 3 | 293 | |||
GenBank mRNA | AF087897.1 | - | 3 | 260 | ||
AF217959.1 | - | 3 | 260 | |||
AF261717.1 | - | 3 | 260 | |||
AF274026.1 | - | 3 | 259 | |||
AF318066.1 | - | 2 | 160 | |||
AK298591.1 | - | 2 | 126 | |||
AK312469.1 | - | 3 | 248 | |||
AL136724.1 | - | 3 | 276 | |||
AY008268.1 | - | 3 | 284 | |||
BC003658.2 | - | 3 | 258 | 3546972 | ||
BG327663.1 | - | 4 | 332 | |||
CR533492.1 | - | 2 | 160 | |||
DB216796.1 | - | 4 | 363 |