Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C8orf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020130.4 | NP_064515.1 | 1 | 200 | ||
GenBank mRNA | AF268037.1 | - | 1 | 184 | ||
AK026240.1 | - | 1 | 192 | |||
BC020623.1 | - | 1 | 186 | 4718061 | ||
BC021672.1 | - | 1 | 204 | 3682270 |