Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168618.1 | NP_001162089.1 | 7 | 859 | ||
NM_020153.3 | NP_064538.3 | 8 | 1012 | |||
XM_011542905.2 | XP_011541207.1 | 8 | 3754 | |||
XM_011542906.2 | XP_011541208.1 | 7 | 3596 | |||
XM_017018017.1 | XP_016873506.1 | 7 | 751 | |||
XM_017018018.1 | XP_016873507.1 | 6 | 722 | |||
GenBank mRNA | AJ249980.1 | - | 7 | 786 | ||
AK025480.1 | - | 8 | 971 | |||
AK289521.1 | - | 7 | 710 | |||
AL136934.1 | - | 7 | 694 | |||
BC011647.2 | - | 7 | 697 | 4300917 | ||
BC012802.2 | - | 7 | 790 | 2822487 | ||
BC022856.1 | - | 7 | 820 | 4078579 | ||
CR457344.1 | - | 5 | 440 |