Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082533.1 | NP_001076002.1 | 6 | 1648 | ||
NM_001082534.1 | NP_001076003.1 | 6 | 1213 | |||
NM_020178.4 | NP_064563.1 | 5 | 1522 | |||
XM_017024878.1 | XP_016880367.1 | 3 | 376 | |||
GenBank mRNA | AB001597.1 | - | 5 | 536 | ||
AB036836.1 | - | 6 | 1438 | |||
AF288385.1 | - | 6 | 1204 | |||
AK294652.1 | - | 4 | 893 | |||
AK313001.1 | - | 5 | 1098 | |||
AL161812.1 | - | 5 | 538 | |||
AY358509.1 | - | 5 | 816 | |||
BC020577.1 | - | 6 | 1180 | 4181377 | ||
BC029865.1 | - | 6 | 1182 | |||
BC047456.1 | - | 5 | 1435 | 5276935 | ||
BC068462.1 | - | 5 | 1481 | 5260321 | ||
CR749607.1 | - | 6 | 984 |