Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020196.2 | NP_064581.2 | 7 | 941 | ||
GenBank mRNA | AB026111.1 | - | 7 | 930 | ||
AB033003.1 | - | 5 | 617 | |||
AF226051.1 | - | 7 | 941 | |||
AF258567.1 | - | 7 | 936 | |||
AF272147.1 | - | 7 | 951 | |||
AK025858.1 | - | 2 | 224 | |||
AK074035.1 | - | 3 | 1512 | |||
BC007208.1 | - | 7 | 923 | 2823235 | ||
BC008778.1 | - | 2 | 256 | |||
CR749864.1 | - | 4 | 470 |