Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other BBX SIRNA Assays ›
Gene Symbol:
BBX
Gene Name:
BBX, HMG-box containing
Gene Aliases:
ARTC1, HBP2, HSPC339, MDS001
Chromosome Location:
Chr.3: 107522936 - 107811329 on Build GRCh38
UniGene ID:
Hs.124366
Species:
Human
siRNA ID
HSS125866
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: BBX
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142568.2
NP_001136040.1
10
1207
NM_001276286.1
NP_001263215.1
10
1207
NM_020235.6
NP_064620.2
10
1207
XM_005247642.3
XP_005247699.1
10
1123
XM_005247643.3
XP_005247700.1
10
1135
XM_005247644.3
XP_005247701.1
11
1298
XM_011512999.2
XP_011511301.1
10
1207
XM_011513000.1
XP_011511302.1
10
1335
XM_011513001.1
XP_011511303.1
10
1175
XM_011513002.2
XP_011511304.1
10
1207
XM_011513003.2
XP_011511305.1
10
1207
XM_011513004.2
XP_011511306.1
10
1335
XM_011513005.2
XP_011511307.1
10
1207
XM_011513006.2
XP_011511308.1
10
1207
XM_011513007.2
XP_011511309.1
10
1207
XM_011513010.2
XP_011511312.1
10
1207
XM_017006881.1
XP_016862370.1
12
1348
XM_017006882.1
XP_016862371.1
11
1298
XM_017006883.1
XP_016862372.1
10
1207
GenBank mRNA
AF161457.1
-
10
1187
AF276948.1
-
7
905
AF276949.1
-
10
1138
AF283775.2
-
10
1201
AF454941.1
-
10
1200
AF454942.1
-
8
1080
AK057010.1
-
10
1153
AK291757.1
-
10
1153
AK307668.1
-
9
1247
AK314434.1
-
10
1158
AL136769.1
-
8
1080
BC110903.1
-
8
1117
BC131718.1
-
10
1100
40123389
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
bone development
DNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-fwm9x:80/10.239.164.221:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0