Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020307.3 | NP_064703.1 | 11 | 2025 | ||
XM_005247648.3 | XP_005247705.1 | 12 | 1482 | |||
XM_006713710.3 | XP_006713773.1 | 10 | 1370 | |||
XM_011513011.2 | XP_011511313.1 | 12 | 2034 | |||
XM_017006891.1 | XP_016862380.1 | 12 | 1588 | |||
XM_017006892.1 | XP_016862381.1 | 12 | 3154 | |||
XM_017006893.1 | XP_016862382.1 | 12 | 3062 | |||
XM_017006894.1 | XP_016862383.1 | 13 | 1619 | |||
XM_017006895.1 | XP_016862384.1 | 13 | 1496 | |||
XM_017006896.1 | XP_016862385.1 | 12 | 1423 | |||
XM_017006897.1 | XP_016862386.1 | 11 | 1458 | |||
GenBank mRNA | AF180920.1 | - | 11 | 1413 | ||
AF208843.1 | - | 7 | 846 | |||
AF367476.1 | - | 13 | 1667 | |||
AK022974.1 | - | 11 | 1412 | |||
AK125606.1 | - | 1 | 1738 | |||
AY358663.1 | - | 6 | 1022 | |||
BC007081.1 | - | 11 | 1412 | 4066376 | ||
BC038394.1 | - | 13 | 1675 | |||
BC067812.1 | - | 11 | 1401 | 5298716 | ||
BX641146.1 | - | 6 | 4132 | |||
BX647554.1 | - | 4 | 3004 |