Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020381.3 | NP_065114.3 | 2 | 598 | ||
XM_011535956.2 | XP_011534258.1 | 2 | 696 | |||
XM_011535957.2 | XP_011534259.1 | 2 | 706 | |||
XM_011535959.2 | XP_011534261.1 | 2 | 671 | |||
XM_011535960.2 | XP_011534262.1 | 2 | 325 | |||
XM_011535961.2 | XP_011534263.1 | 2 | 618 | |||
XM_011535962.2 | XP_011534264.1 | 2 | 240 | |||
XM_011535963.2 | XP_011534265.1 | 2 | 618 | |||
XM_017011082.1 | XP_016866571.1 | 2 | 618 | |||
GenBank mRNA | AB210839.1 | - | 2 | 308 | ||
AF254956.1 | - | 2 | 569 | |||
AK225629.1 | - | 2 | 579 | |||
AK296717.1 | - | 2 | 579 | |||
AK301480.1 | - | 2 | 201 | |||
AK315804.1 | - | 2 | 579 | |||
BC029491.1 | - | 2 | 588 | |||
BC039906.1 | - | 2 | 584 | 6066481 |