Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDSS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020381.3 | NP_065114.3 | 5 | 1018 | ||
XM_011535956.2 | XP_011534258.1 | 5 | 1116 | |||
XM_011535957.2 | XP_011534259.1 | 5 | 1126 | |||
XM_011535958.2 | XP_011534260.1 | 4 | 982 | |||
XM_011535959.2 | XP_011534261.1 | 5 | 1091 | |||
XM_011535960.2 | XP_011534262.1 | 5 | 745 | |||
XM_011535962.2 | XP_011534264.1 | 5 | 660 | |||
XM_017011082.1 | XP_016866571.1 | 5 | 1038 | |||
GenBank mRNA | AB210839.1 | - | 5 | 728 | ||
AF254956.1 | - | 5 | 989 | |||
AK225629.1 | - | 5 | 999 | |||
AK301480.1 | - | 5 | 621 | |||
AK315804.1 | - | 5 | 999 | |||
BC039906.1 | - | 5 | 1004 | 6066481 |