Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020387.3 | NP_065120.2 | 2 | 332 | ||
GenBank mRNA | AF083124.1 | - | 2 | 293 | ||
AF274025.1 | - | 2 | 311 | |||
BC009831.2 | - | 2 | 306 | 3926839 | ||
BC033322.1 | - | 2 | 296 | 4775725 | ||
BP263750.1 | - | 2 | 332 |