Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAN1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289010.1 | NP_001275939.1 | 3 | 992 | ||
NM_020379.3 | NP_065112.1 | 3 | 992 | |||
GenBank mRNA | AB209275.1 | - | 3 | 340 | ||
AF261655.1 | - | 3 | 992 | |||
AK128560.1 | - | 2 | 60 | |||
BC137017.1 | - | 3 | 798 | 9021010 |