Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RTN4 SIRNA Assays ›
Gene Symbol:
RTN4
Gene Name:
reticulon 4
Gene Aliases:
ASY, NI220/250, NOGO, NOGO-A, NOGOC, NSP, NSP-CL, Nbla00271, Nbla10545, Nogo-B, Nogo-C, RTN-X, RTN4-A, RTN4-B1, RTN4-B2, RTN4-C
Chromosome Location:
Chr.2: 54972187 - 55137831 on Build GRCh38
UniGene ID:
Hs.637850
Species:
Human
siRNA ID
HSS126049
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RTN4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001321859.1
NP_001308788.1
4
2575
NM_001321860.1
NP_001308789.1
4
2645
NM_001321861.1
NP_001308790.1
4
2608
NM_001321862.1
NP_001308791.1
4
2595
NM_001321863.1
NP_001308792.1
4
2545
NM_001321904.1
NP_001308833.1
5
3023
NM_007008.2
NP_008939.1
2
265
NM_020532.4
NP_065393.1
4
3328
NM_153828.2
NP_722550.1
2
871
NM_207520.1
NP_997403.1
3
928
NM_207521.1
NP_997404.1
4
2586
NR_135829.1
2
151
NR_135830.1
2
129
XM_005264434.3
XP_005264491.1
5
2722
XM_017004518.1
XP_016860007.1
5
2761
XM_017004519.1
XP_016860008.1
5
2878
GenBank mRNA
AB015639.1
-
2
639
AB020693.1
-
4
3164
AB040462.1
-
4
3274
AB040463.1
-
2
817
AB073351.1
-
2
1000
AF063601.1
-
2
1588
AF077050.1
-
2
264
AF087901.1
-
2
156
AF125103.1
-
2
265
AF132047.1
-
2
752
AF132048.1
-
2
171
AF148537.1
-
4
3171
AF148538.1
-
2
714
AF177332.1
-
2
170
AF320999.1
-
3
2334
AF333336.1
-
4
2590
AJ251383.1
-
4
3030
AJ251384.1
-
2
573
AJ251385.1
-
2
51
AK075039.1
-
2
805
AK129806.1
-
2
730
AK130812.1
-
2
632
AY102276.1
-
2
285
AY102277.1
-
2
817
AY102278.1
-
3
874
AY102279.1
-
4
3274
AY123245.1
-
4
2587
AY123246.1
-
4
2645
AY123247.1
-
4
2608
AY123248.1
-
4
2594
AY123249.1
-
4
2555
AY123250.1
-
4
2545
BC001035.2
-
2
236
BC007109.1
-
2
162
BC010737.1
-
2
578
BC012619.1
-
2
728
BC014366.1
-
2
162
BC016165.1
-
3
877
BC026788.1
-
2
759
BC068991.1
-
2
821
BC071848.1
-
3
559
BC150182.1
-
4
3278
40148210
BC152425.1
-
4
3164
8862539
BC152555.1
-
4
3278
40148984
CN429712.1
-
2
100
DQ778739.1
-
2
51
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
apoptotic process
axonal fasciculation
cerebral cortex radial glia guided migration
negative regulation of cell growth
negative regulation of axon extension
regulation of apoptotic process
negative regulation of axonogenesis
nuclear pore complex assembly
cardiac epithelial to mesenchymal transition
endoplasmic reticulum tubular network organization
endoplasmic reticulum tubular network assembly
cell-cell adhesion
regulation of branching morphogenesis of a nerve
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-x5whk:80/10.239.163.84:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0