Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142545.1 | NP_001136017.1 | 4 | 462 | ||
NM_020421.3 | NP_065154.2 | 5 | 666 | |||
XM_005267885.3 | XP_005267942.1 | 5 | 689 | |||
XM_005267886.3 | XP_005267943.1 | 5 | 685 | |||
XM_011536987.2 | XP_011535289.1 | 5 | 685 | |||
XM_011536988.2 | XP_011535290.1 | 5 | 685 | |||
XM_017021474.1 | XP_016876963.1 | 5 | 685 | |||
XM_017021475.1 | XP_016876964.1 | 5 | 685 | |||
XM_017021476.1 | XP_016876965.1 | 6 | 647 | |||
XM_017021477.1 | XP_016876966.1 | 3 | 364 | |||
GenBank mRNA | AK096919.1 | - | 4 | 408 | ||
BC017459.1 | - | 5 | 623 | |||
BC058906.1 | - | 5 | 618 | 6150933 | ||
BX248292.1 | - | 5 | 649 | |||
DN996748.1 | - | 4 | 416 |