Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032546.3 | NP_115935.3 | 1 | 363 | ||
NM_187841.2 | NP_912730.2 | 1 | 363 | |||
GenBank mRNA | AJ291714.1 | - | 1 | 60 | ||
AK300699.1 | - | 1 | 364 | |||
BC141807.1 | - | 1 | 135 | 40147443 | ||
DA897004.1 | - | 1 | 363 |