Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNFX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021035.2 | NP_066363.1 | 14 | 4859 | ||
XM_006723836.1 | XP_006723899.1 | 14 | 4689 | |||
XM_011528924.1 | XP_011527226.1 | 13 | 4734 | |||
XM_017027971.1 | XP_016883460.1 | 12 | 3785 | |||
GenBank mRNA | AB037825.2 | - | 14 | 4677 | ||
AK023836.1 | - | 10 | 2611 | |||
BC062347.1 | - | 1 | 198 |