Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288951.1 | NP_001275880.1 | 18 | 2405 | ||
NM_001288953.1 | NP_001275882.1 | 18 | 2324 | |||
NM_001288955.1 | NP_001275884.1 | 16 | 2175 | |||
NM_020458.3 | NP_065191.2 | 17 | 2333 | |||
XM_005264439.3 | XP_005264496.1 | 18 | 2486 | |||
XM_011532998.2 | XP_011531300.1 | 19 | 2551 | |||
XM_011532999.2 | XP_011531301.1 | 17 | 2276 | |||
XM_011533000.2 | XP_011531302.1 | 14 | 1408 | |||
XM_011533001.2 | XP_011531303.1 | 10 | 1198 | |||
XM_017004524.1 | XP_016860013.1 | 16 | 2160 | |||
XM_017004525.1 | XP_016860014.1 | 17 | 1818 | |||
XM_017004526.1 | XP_016860015.1 | 14 | 2028 | |||
XM_017004527.1 | XP_016860016.1 | 17 | 2428 | |||
XM_017004528.1 | XP_016860017.1 | 18 | 2493 | |||
XM_017004529.1 | XP_016860018.1 | 17 | 2276 | |||
GenBank mRNA | AB032966.2 | - | 18 | 1913 | ||
AK056464.1 | - | 18 | 2296 | |||
AL834383.1 | - | 17 | 2333 | |||
BC001978.1 | - | 8 | 742 | |||
BC065554.1 | - | 14 | 1451 | |||
BC082965.1 | - | 9 | 1072 | |||
BC111487.1 | - | 18 | 2293 | 6041916 | ||
BC114365.2 | - | 16 | 1806 | 40038218 |