Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020673.2 | NP_065724.1 | 5 | 592 | ||
GenBank mRNA | AF091034.2 | - | 5 | 570 | ||
AJ276210.1 | - | 5 | 557 | |||
AK091180.1 | - | 5 | 346 | |||
BC015710.1 | - | 5 | 517 | 3907891 | ||
BC063457.1 | - | 5 | 576 | 5182786 | ||
BT007046.1 | - | 5 | 330 |