Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPC25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020675.3 | NP_065726.1 | 6, 7 | 676 | ||
GenBank mRNA | AF225416.1 | - | 6, 7 | 658 | ||
AK291093.1 | - | 6, 7 | 687 | |||
AY456388.1 | - | 5, 6 | 545 | |||
BC022255.1 | - | 6, 7 | 676 | 4738187 | ||
CR457197.1 | - | 5, 6 | 545 |