Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARRDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020801.2 | NP_065852.1 | 1, 2 | 518 | ||
GenBank mRNA | AB037797.1 | - | 1, 2 | 490 | ||
AK291753.1 | - | 1, 2 | 493 | |||
BC015928.2 | - | 1, 2 | 485 | 3908916 | ||
BC053619.1 | - | 1, 2 | 490 | 6155815 | ||
DA110572.1 | - | 1, 2 | 518 |