Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIP2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173602.2 | NP_775873.2 | 13 | 1790 | ||
XM_005269044.1 | XP_005269101.1 | 13 | 1793 | |||
XM_006719520.3 | XP_006719583.1 | 14 | 1766 | |||
XM_006719521.3 | XP_006719584.1 | 13 | 1603 | |||
XM_011538594.2 | XP_011536896.1 | 14 | 1819 | |||
XM_011538596.2 | XP_011536898.1 | 13 | 1606 | |||
XM_017019699.1 | XP_016875188.1 | 14 | 1801 | |||
XM_017019700.1 | XP_016875189.1 | 13 | 1775 | |||
XM_017019701.1 | XP_016875190.1 | 13 | 1772 | |||
XM_017019703.1 | XP_016875192.1 | 12 | 1569 | |||
XM_017019704.1 | XP_016875193.1 | 12 | 1577 | |||
XM_017019705.1 | XP_016875194.1 | 12 | 1559 | |||
GenBank mRNA | AB040896.2 | - | 4 | 404 |