Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RANBP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320238.1 | NP_001307167.1 | 7, 8 | 1003 | ||
NM_001320239.1 | NP_001307168.1 | 7, 8 | 947 | |||
NM_001320240.1 | NP_001307169.1 | 11, 12 | 1568 | |||
NM_020850.2 | NP_065901.1 | 8, 9 | 1115 | |||
XM_005256070.2 | XP_005256127.1 | 8, 9 | 1023 | |||
XM_005256072.2 | XP_005256129.1 | 7, 8 | 970 | |||
XM_006721238.3 | XP_006721301.1 | 3, 4 | 780 | |||
XM_011523245.2 | XP_011521547.1 | 5, 6 | 762 | |||
XM_011523247.2 | XP_011521549.1 | 5, 6 | 568 | |||
XM_017023507.1 | XP_016878996.1 | 5, 6 | 629 | |||
XM_017023508.1 | XP_016878997.1 | 9, 10 | 1183 | |||
XM_017023509.1 | XP_016878998.1 | 6, 7 | 618 | |||
GenBank mRNA | AB040897.1 | - | 8, 9 | 1004 | ||
AK000686.1 | - | 1246 | ||||
AK055727.1 | - | 11, 12 | 1473 | |||
AK295530.1 | - | 7, 8 | 911 | |||
AK298806.1 | - | 7, 8 | 874 | |||
AY337313.1 | - | 8, 9 | 1115 | |||
BC099917.1 | - | 8, 9 | 1020 | 30528481 | ||
BC121176.1 | - | 8, 9 | 1113 | 40122561 | ||
BC121177.2 | - | 8, 9 | 1113 | 40122562 |