Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DPP10 SIRNA Assays ›
Gene Symbol:
DPP10
Gene Name:
dipeptidyl peptidase like 10
Gene Aliases:
DPL2, DPPY, DPRP-3, DPRP3
Chromosome Location:
Chr.2: 114442358 - 115845752 on Build GRCh38
UniGene ID:
Hs.580539
Species:
Human
siRNA ID
HSS126572
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: DPP10
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001004360.3
NP_001004360.2
20
1921
NM_001178034.1
NP_001171505.1
20
1848
NM_001178036.1
NP_001171507.1
21
2325
NM_001178037.1
NP_001171508.1
20
1891
NM_001321905.1
NP_001308834.1
21
2351
NM_001321906.1
NP_001308835.1
21
1967
NM_001321907.1
NP_001308836.1
19
2197
NM_001321908.1
NP_001308837.1
20
2210
NM_001321909.1
NP_001308838.1
19
1825
NM_001321910.1
NP_001308839.1
21
2294
NM_001321911.1
NP_001308840.1
20
2293
NM_001321912.1
NP_001308841.1
21
2325
NM_001321913.1
NP_001308842.1
19
2121
NM_001321914.1
NP_001308843.1
20
1846
NM_020868.4
NP_065919.2
20
2236
XM_017004565.1
XP_016860054.1
20
2356
XM_017004566.1
XP_016860055.1
19
1840
GenBank mRNA
AB040925.2
-
20
1803
AK025075.1
-
8
647
AK289967.1
-
20
1935
AK296490.1
-
20
1923
AK315147.1
-
20
2040
AY172661.3
-
20
1815
AY387785.1
-
20
1837
BC030832.1
-
20
2272
4811771
DQ857321.1
-
20
1803
DQ857322.1
-
20
1827
DQ857323.1
-
20
1794
Back To Top
More Information
Panther Classification:
Molecular Function -
serine protease
Gene Ontology Categories:
Function(s)
Process(es)
proteolysis
protein localization to plasma membrane
positive regulation of establishment of protein localization to plasma membrane
regulation of potassium ion transmembrane transport
serine-type peptidase activity
dipeptidyl-peptidase activity
potassium channel regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-jmrqp:80/10.239.165.107:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0