Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020893.2 | NP_065944.2 | 24 | 3497 | ||
GenBank mRNA | CR749320.1 | - | 24 | 3439 |
Gene Symbol: LOC100499484-C9ORF174 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036527.1 | 35 | 4288 | |||
NR_036528.1 | 38 | 4792 | ||||
NR_036529.1 | 32 | 3848 |