Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAT1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020927.2 | NP_065978.1 | 1, 2 | 358 | ||
GenBank mRNA | AB046796.1 | - | 1, 2 | 328 | ||
AK290153.1 | - | 1, 2 | 336 | |||
BC033537.1 | - | 1, 2 | 336 | 4821265 | ||
HY132852.1 | - | 1, 2 | 359 |