Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020943.2 | NP_065994.1 | 20 | 2478 | ||
XM_005246726.2 | XP_005246783.1 | 20 | 2581 | |||
GenBank mRNA | AB046824.1 | - | 19 | 2297 | ||
AK025635.1 | - | 20 | 2334 | |||
AK026978.1 | - | 13 | 1435 | |||
AK123647.1 | - | 7 | 1435 | |||
BC016651.1 | - | 19 | 2302 | |||
BC031216.1 | - | 20 | 2478 | |||
BC041662.1 | - | 13 | 1429 | |||
BC053573.1 | - | 17 | 2000 | |||
BC057826.1 | - | 20 | 2336 | |||
BC093952.1 | - | 19 | 2200 | 7939797 | ||
BC093954.1 | - | 19 | 2200 | 7939799 | ||
BC143433.1 | - | 16 | 1960 |