Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MIER1 SIRNA Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
UniGene ID:
Hs.605432
Species:
Human
siRNA ID
HSS126700
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1299001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MIER1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001077700.2
NP_001071168.2
11
1152
NM_001077701.2
NP_001071169.1
10
1008
NM_001077702.2
NP_001071170.2
12
1226
NM_001077703.2
NP_001071171.2
11
1152
NM_001077704.2
NP_001071172.1
10
1008
NM_001146110.1
NP_001139582.1
12
1229
NM_001146111.1
NP_001139583.1
12
1226
NM_001146112.1
NP_001139584.1
12
1187
NM_001146113.1
NP_001139585.1
10
1006
NM_020948.3
NP_065999.2
12
1226
XM_005271076.3
XP_005271133.1
12
1264
XM_011541864.2
XP_011540166.1
13
1171
XM_011541865.2
XP_011540167.1
13
1132
XM_017001919.1
XP_016857408.1
13
1148
XM_017001920.1
XP_016857409.1
13
1165
XM_017001921.1
XP_016857410.1
13
1193
XM_017001922.1
XP_016857411.1
13
1168
XM_017001923.1
XP_016857412.1
11
988
XM_017001924.1
XP_016857413.1
12
1120
XM_017001925.1
XP_016857414.1
12
1225
XM_017001926.1
XP_016857415.1
13
1164
XM_017001927.1
XP_016857416.1
11
1037
XM_017001928.1
XP_016857417.1
11
1113
XM_017001929.1
XP_016857418.1
12
1094
XM_017001930.1
XP_016857419.1
11
1038
XM_017001931.1
XP_016857420.1
12
1072
XM_017001932.1
XP_016857421.1
10
1001
XM_017001933.1
XP_016857422.1
9
855
XM_017001934.1
XP_016857423.1
8
751
GenBank mRNA
AB046830.1
-
8
777
AF515446.1
-
10
1008
AF515447.1
-
11
1089
AF515448.2
-
12
1163
AK302061.1
-
11
1152
AK307717.1
-
12
1123
AL831987.2
-
12
1168
AY124186.2
-
12
1163
AY124187.1
-
11
1089
AY124188.1
-
10
1008
AY124189.2
-
12
1163
AY124190.1
-
11
1089
AY124191.1
-
10
1008
AY124192.2
-
12
1163
AY124193.1
-
11
1089
AY124194.1
-
10
1008
BC108726.1
-
12
1107
5194296
BC125217.1
-
9
804
40123797
BC125218.1
-
9
804
40123803
CR627441.1
-
12
1063
Back To Top
More Information
Panther Classification:
Molecular Function -
homeodomain transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0