Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EPG5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020964.2 | NP_066015.2 | 41 | 7264 | ||
XM_011526122.1 | XP_011524424.1 | 41 | 7264 | |||
XM_017025889.1 | XP_016881378.1 | 41 | 7243 | |||
XM_017025890.1 | XP_016881379.1 | 41 | 7246 | |||
XM_017025891.1 | XP_016881380.1 | 40 | 7123 | |||
XM_017025892.1 | XP_016881381.1 | 40 | 7289 | |||
XM_017025893.1 | XP_016881382.1 | 33 | 5136 | |||
GenBank mRNA | AK023817.1 | - | 25 | 3943 | ||
AL583915.1 | - | 2 | 273 | |||
BC003695.1 | - | 5 | 943 | |||
BC036911.1 | - | 5 | 950 | |||
BC130614.1 | - | 24 | 3797 | |||
BC144104.1 | - | 22 | 3458 | |||
BC144105.1 | - | 23 | 3690 |