Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB40C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172663.1 | NP_001166134.1 | 6 | 663 | ||
NM_001172664.1 | NP_001166135.1 | 6 | 555 | |||
NM_001172665.1 | NP_001166136.1 | 6 | 515 | |||
NM_001172666.1 | NP_001166137.1 | 6 | 458 | |||
NM_021168.4 | NP_066991.3 | 5 | 560 | |||
GenBank mRNA | AY823398.1 | - | 5 | 428 | ||
BC001581.1 | - | 2 | 154 | |||
BC028696.1 | - | 5 | 562 | 4826800 | ||
BG696681.1 | - | 6 | 664 | |||
BM545442.1 | - | 5 | 634 | |||
BQ685517.1 | - | 6 | 494 | |||
BX442451.2 | - | 6 | 604 |