Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 9 | 1148 | ||
NR_033902.1 | 9 | 966 | ||||
XM_011544603.1 | XP_011542905.1 | 9 | 966 | |||
XM_011544604.1 | XP_011542906.1 | 8 | 846 | |||
XM_017013717.1 | XP_016869206.1 | 9 | 1148 | |||
GenBank mRNA | AB075847.1 | - | 6 | 457 | ||
AK095712.1 | - | 9 | 822 | |||
AK314535.1 | - | 9 | 841 | |||
AL834162.1 | - | 9 | 1135 | |||
AL834351.1 | - | 9 | 1136 | |||
AL834352.1 | - | 9 | 1136 | |||
BC065495.1 | - | 9 | 824 | 5496068 | ||
BX390251.1 | - | 4 | 332 | |||
BX640952.1 | - | 9 | 835 |