Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021174.5 | NP_066997.3 | 4 | 598 | ||
NR_033902.1 | 4 | 416 | ||||
XM_011544603.1 | XP_011542905.1 | 4 | 416 | |||
XM_011544604.1 | XP_011542906.1 | 4 | 416 | |||
XM_017013717.1 | XP_016869206.1 | 4 | 598 | |||
GenBank mRNA | AK095712.1 | - | 4 | 272 | ||
AK314535.1 | - | 4 | 291 | |||
AL834162.1 | - | 4 | 585 | |||
AL834351.1 | - | 4 | 586 | |||
AL834352.1 | - | 4 | 586 | |||
BC065495.1 | - | 4 | 274 | 5496068 | ||
BX640952.1 | - | 4 | 285 | |||
DA721640.1 | - | 4 | 416 |