Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNB1IP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021178.4 | NP_067001.3 | 6, 7 | 1123 | ||
NM_182849.2 | NP_878269.1 | 5, 6 | 923 | |||
NM_182852.3 | NP_878272.1 | 7, 8 | 1304 | |||
GenBank mRNA | AA101731.1 | - | 1, 2 | 379 | ||
AF216381.1 | - | 2, 3 | 634 | |||
AI301662.1 | - | 1, 2 | 381 | |||
AK026233.1 | - | 6, 7 | 1090 | |||
AK225253.1 | - | 6, 7 | 1095 | |||
AK225451.1 | - | 6, 7 | 1090 | |||
AK226037.1 | - | 6, 7 | 1095 | |||
AL161994.1 | - | 6, 7 | 1103 | |||
BC000369.2 | - | 5, 6 | 1065 | 2819346 | ||
BC001218.1 | - | 5, 6 | 1068 | 3528496 | ||
BC004435.1 | - | 5, 6 | 1065 | 2819346 | ||
BX161423.1 | - | 6, 7 | 1283 | |||
BX161424.1 | - | 6, 7 | 1251 | |||
BX247978.1 | - | 6, 7 | 1231 |