Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00474 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024032.1 | 2 | 236 | |||
GenBank mRNA | AB021923.1 | - | 2 | 345 | ||
BC104240.1 | - | 2 | 236 | |||
BC104241.1 | - | 2 | 235 | |||
BC112996.1 | - | 2 | 236 | |||
BC112997.1 | - | 2 | 236 | |||
BC120871.2 | - | 2 | 332 | |||
BC128039.1 | - | 2 | 236 |