Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RBM25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021239.2 | NP_067062.1 | 6 | 659 | ||
XM_011537044.2 | XP_011535346.1 | 7 | 761 | |||
XM_011537045.2 | XP_011535347.1 | 5 | 466 | |||
GenBank mRNA | AK026107.1 | - | 6 | 639 | ||
AK094697.1 | - | 4 | 398 | |||
AL832314.1 | - | 6 | 630 | |||
BC062440.1 | - | 6 | 663 | |||
BC083496.1 | - | 6 | 625 | |||
BC111066.1 | - | 6 | 623 | |||
BC113389.1 | - | 5 | 439 | |||
BC113391.1 | - | 5 | 439 | |||
BC136775.1 | - | 6 | 496 | 9020765 | ||
BC136776.1 | - | 6 | 496 | 9020766 | ||
BC144407.1 | - | 7 | 564 | 9052928 | ||
BX647116.1 | - | 6 | 612 |