Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CACNG7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031896.4 | NP_114102.2 | 2 | 346 | ||
XM_005259124.3 | XP_005259181.1 | 3 | 373 | |||
XM_017027093.1 | XP_016882582.1 | 3 | 373 | |||
GenBank mRNA | AF288387.1 | - | 2 | 266 | ||
AF361353.1 | - | 2 | 251 | |||
AF458897.1 | - | 2 | 251 | |||
BC069332.1 | - | 2 | 266 | 7262204 | ||
BC093869.1 | - | 2 | 346 | 7939714 | ||
BC113503.1 | - | 2 | 346 | 8322555 |