Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136039.2 | NP_001129511.1 | 4 | 778 | ||
NM_001142355.1 | NP_001135827.1 | 4 | 727 | |||
NM_001142356.1 | NP_001135828.1 | 3 | 713 | |||
NM_021824.3 | NP_068596.2 | 4 | 930 | |||
XM_005246742.3 | XP_005246799.1 | 4 | 993 | |||
XM_011511582.1 | XP_011509884.1 | 4 | 914 | |||
XM_011511583.1 | XP_011509885.1 | 4 | 1658 | |||
XM_011511584.1 | XP_011509886.1 | 4 | 1476 | |||
XM_011511585.1 | XP_011509887.1 | 4 | 1425 | |||
XM_017004630.1 | XP_016860119.1 | 4 | 1175 | |||
GenBank mRNA | AB038949.1 | - | 4 | 891 | ||
AF060513.1 | - | 4 | 687 | |||
AF182416.1 | - | 4 | 852 | |||
AF283538.1 | - | 4 | 733 | |||
AK023378.1 | - | 4 | 876 | |||
AK225905.1 | - | 4 | 2253 | |||
AK314709.1 | - | 4 | 732 | |||
AL834430.1 | - | 3 | 553 | |||
AY251943.1 | - | 3 | 716 | |||
AY251944.1 | - | 3, 4 | 716 | |||
BC007654.2 | - | 3 | 714 | 3346099 |