Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021942.5 | NP_068761.4 | 26 | 3135 | ||
NM_199053.2 | NP_951008.1 | 26 | 3135 | |||
XM_017008537.1 | XP_016864026.1 | 26 | 3135 | |||
XM_017008539.1 | XP_016864028.1 | 24 | 2786 | |||
GenBank mRNA | AK022778.1 | - | 5 | 539 | ||
AK023390.1 | - | 7 | 824 | |||
AK090489.1 | - | 1 | 926 | |||
AK091146.1 | - | 21 | 2359 | |||
AK096345.1 | - | 26 | 3109 | |||
AK315057.1 | - | 21 | 2341 | |||
AL136752.1 | - | 18 | 2044 | |||
AL833451.1 | - | 25 | 2941 | |||
BC051724.1 | - | 26 | 3079 | 5265781 | ||
BC139745.1 | - | 14 | 1515 | |||
BX538044.1 | - | 26 | 3089 | |||
BX647127.1 | - | 26 | 3106 | |||
BX648499.1 | - | 26 | 3110 | |||
CR533553.1 | - | 18 | 1979 |