Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022061.3 | NP_071344.1 | 3 | 436 | ||
GenBank mRNA | AF141338.1 | - | 3 | 358 | ||
AF164797.1 | - | 3 | 351 | |||
AK026857.1 | - | 3 | 341 | |||
BC012306.1 | - | 3 | 337 | 4706168 | ||
BQ066780.1 | - | 378 | ||||
CR457177.1 | - | 3 | 331 | |||
DB228622.1 | - | 3 | 436 |